Learn More
Medchemexpress LLC 2,4,6-trimethylbenzaldehyde | 487-68-3 | MFCD00003341 | 99.2% | 148.20 g/mol | C10H12O | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0174691KG
Encompass_Preferred
Mesitaldehyde (2,4,6-trimethylbenzaldehyde, CAS 487-68-3) is an aromatic aldehyde supplied as a high-purity research reagent for organic synthesis and analytical applications. It has formula C10H12O and molecular weight 148.20 g/mol, and is certified at 99.15% purity per the certificate of analysis.
  • High purity suitable for research and synthesis.
  • Verified identity with batch-specific certificate of analysis available.
  • Useful as an intermediate in organic synthesis and as an analytical reference standard.
  • Aromatic aldehyde with formula C10H12O and molecular weight 148.20 g/mol.
Catalog No. 50-003-00412