Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylbenzenesulfonyl hydrazide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
16182-15-3
C9H14N2O2S
214.283
MFCD00007585
JQUBKTQDNVZHIY-UHFFFAOYSA-N
2,4,6-trimethylbenzenesulfonyl hydrazide,mesitylene-2-sulphonohydrazide,2-mesitylenesulfonyl hydrazide,2-mesitylenesulfonohydrazide,2-mesitylenesulfono hydrazide,benzenesulfonic acid,2,4,6-trimethyl-, hydrazide,hydrazino 2,4,6-trimethylphenyl sulfone,mesitylenesulphonohydrazide,mesitylenesulfonylhydrazide,2-mesitylehesulfonohydrazide
85321
2,4,6-trimethylbenzenesulfonohydrazide
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C
16182-15-3
C9H14N2O2S
214.283
MFCD00007585
JQUBKTQDNVZHIY-UHFFFAOYSA-N
2,4,6-trimethylbenzenesulfonyl hydrazide,mesitylene-2-sulphonohydrazide,2-mesitylenesulfonyl hydrazide,2-mesitylenesulfonohydrazide,2-mesitylenesulfono hydrazide,benzenesulfonic acid,2,4,6-trimethyl-, hydrazide,hydrazino 2,4,6-trimethylphenyl sulfone,mesitylenesulphonohydrazide,mesitylenesulfonylhydrazide,2-mesitylehesulfonohydrazide
85321
2,4,6-trimethylbenzenesulfonohydrazide
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C
Packaging Glass Bottle
Infrared Spectrum Authentic
Color White
Melting Point 110.0°C to 114.0°C
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)3C6H2SO2NHNH2
Formula Weight 214.28
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Trimethylbenzenesulfonyl hydrazide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 240-319-4

TSCA : TSCA