Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylbenzyl alcohol, 98+%

Catalog No. AAL0195206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0195206 5 g
AAL0195214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0195206 Supplier Thermo Scientific Chemicals Supplier No. L0195206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4170-90-5 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4170-90-5
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00014422
InChI Key LODDFDHPSIYCTK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl alcohol, mesitylmethanol, mesitylcarbinol, 2,4,6-trimethylphenyl methanol, unii-rev3kvt84n, benzenemethanol, 2,4,6-trimethyl, 2,4,6-trimethylbenzylic alcohol, benzyl alcohol, 2,4,6-trimethyl, rev3kvt84n, 2,4,6-trimethylphenyl methan-1-ol
PubChem CID 20139
IUPAC Name (2,4,6-trimethylphenyl)methanol
SMILES CC1=CC(=C(C(=C1)C)CO)C

Specifications

Melting Point 85°C to 89°C
Boiling Point 140°C (15 mmHg)
Quantity 5 g
Beilstein 1862608
Formula Weight 150.22
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trimethylbenzyl alcohol
EINECSNumber 224-032-1
RTECSNumber DP1402000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.