Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4,6-trimethylphenol | 527-60-6 | MFCD00002235 | 100.0% | 136.19 g/mol | C9H12O | 25 G
SDP

Catalog No. 5000303711
Encompass_Preferred

2,4,6-Trimethylphenol (mesitol) is an aromatic phenol used as an analytical standard and probe compound. It reacts mainly with dissolved organic matter and is rapidly oxidized by singlet oxygen in aqueous solution, making it useful in photochemical and oxidation studies and as a reference material for analytical methods.

  • Chemical formula: C9H12O.
  • Molecular weight: 136.19 g/mol.
  • CAS number: 527-60-6.
  • High purity (reported 65;99.48%).
  • Available in multiple solid and solution pack sizes for analytical use.
  • Suitable as a probe in photochemical and oxidation experiments.

Catalog No. 50-003-03711 Supplier Medchemexpress LLC Supplier No. 25GHYW038786
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4,6-Trimethylphenol (mesitol) is an aromatic phenol used as an analytical standard and probe compound. It reacts mainly with dissolved organic matter and is rapidly oxidized by singlet oxygen in aqueous solution, making it useful in photochemical and oxidation studies and as a reference material for analytical methods.

  • Chemical formula: C9H12O.
  • Molecular weight: 136.19 g/mol.
  • CAS number: 527-60-6.
  • High purity (reported 65;99.48%).
  • Available in multiple solid and solution pack sizes for analytical use.
  • Suitable as a probe in photochemical and oxidation experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.