Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4,6-trimethylphenol | 527-60-6 | MFCD00002235 | 99.5% | 136.19 g/mol | C9H12O | 500 MG
SDP

Catalog No. 5000301167
Encompass_Preferred

2,4,6-Trimethylphenol is an aromatic phenol used as a probe compound in research; it reacts mainly with organic matter (3DOM) and is rapidly oxidized by singlet oxygen in aqueous solution. For research use only.

  • Probe compound for studies of organic matter reactivity (3DOM).
  • Undergoes rapid oxidation by singlet oxygen in aqueous environments.
  • High reported purity: 99.48% (data sheet) and 99.98% (documentation).
  • Molecular formula C9H12O; molecular weight 136.19 g/mol.
  • Solubility: DMSO 100 mg/mL (in vitro); formulatable for in vivo dosing at ≥ 2.5 mg/mL in various vehicles.
  • Storage: store at 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-01167 Supplier Medchemexpress LLC Supplier No. HYW038786500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4,6-Trimethylphenol is an aromatic phenol used as a probe compound in research; it reacts mainly with organic matter (3DOM) and is rapidly oxidized by singlet oxygen in aqueous solution. For research use only.

  • Probe compound for studies of organic matter reactivity (3DOM).
  • Undergoes rapid oxidation by singlet oxygen in aqueous environments.
  • High reported purity: 99.48% (data sheet) and 99.98% (documentation).
  • Molecular formula C9H12O; molecular weight 136.19 g/mol.
  • Solubility: DMSO 100 mg/mL (in vitro); formulatable for in vivo dosing at ≥ 2.5 mg/mL in various vehicles.
  • Storage: store at 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.