Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylpyridine 98.0+%, TCI America™
SDP

Catalog No. T0716100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0716100ML 100 mL
T071625ML 25 mL
T0716500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0716100ML Supplier TCI America Supplier No. T0716100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-75-8
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00006338
InChI Key BWZVCCNYKMEVEX-UHFFFAOYSA-N
Synonym 2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
PubChem CID 7953
IUPAC Name 2,4,6-trimethylpyridine
SMILES CC1=CC(C)=NC(C)=C1

Specifications

Melting Point -43°C
Boiling Point 171°C
Quantity 100 mL
UN Number 1992
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylpyridine
EINECSNumber (5)-0715
RTECSNumber UU0970000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.