Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylpyrylium tetrafluoroborate, 98+%

Catalog No. AAL0046706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0046706 5 g
AAL0046714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0046706 Supplier Thermo Scientific Chemicals Supplier No. L0046706
  • $84.20
    $71.65 / Each of 1

    Save $12.55

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 773-01-3 | C8H11BF4O | 209.98 g/mol

It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.

Solubility
Soluble in water and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-01-3
Molecular Formula C8H11BF4O
Molecular Weight (g/mol) 209.98
MDL Number MFCD00032098
InChI Key PYOPRIFMELCENN-UHFFFAOYSA-N
Synonym 2,4,6-trimethylpyrylium tetrafluoroborate, 2,4,6-trimethyl-1??-pyran-1-ylium tetrafluoroborate, 2,4,6-trimethylpyrylium tetrafluoborate, 2,4,6-trimethylpyryliumtetrafluoroborate, pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-, pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-1:1, 2,4,6-trimethyl-1$l^ 4-pyran-1-ylium tetrafluoroborate
PubChem CID 164732
IUPAC Name 2,4,6-trimethylpyrylium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC1=CC(C)=[O+]C(C)=C1

Specifications

Melting Point 224°C to 226°C
Quantity 5 g
Beilstein 3639361
Solubility Information Soluble in water and methanol.
Formula Weight 209.98
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trimethylpyrylium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-256-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.