Learn More
Strem, An Ascensus Company CAS# 68957-94-8. 10g. 2,4,6-Tripropyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane T3P (Propylphosphonic acid anhydride 50% solution in ethyl acetate). MFCD00006583
SDP
Supplier:  Strem, An Ascensus Company 15916010G
Encompass
CAS# 68957-94-8. 10g. 2,4,6-Tripropyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane T3P (Propylphosphonic acid anhydride 50% solution in ethyl acetate). MFCD00006583. Molecular Weight: 318.20. Molecular Formula: (C3H7O2P)3. Color/form: slightly yellow to brown liq. Strem# 15-9160. http://www.strem.com/catalog/v/15-9160/
Catalog No. 50-371-8