Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4,6-Tris(dimethylaminomethyl)phenol

Catalog No. T5820350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T5820350ML 50 mL
T58203250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T5820350ML Supplier Sigma Aldrich Supplier No. T5820350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,4,6-tris(dimethylaminomethyl)phenol can be used:
  • In the synthesis of water-soluble metal phthalocyanines bearing twelve dimethylamino groups
  • To prepare phenolate anion-based branched/cross-linked anion exchange membranes (aems)
  • As a curing agent in the production of epoxy membranes

Specifications

CAS 90-72-2
Density 0.969 g/mL (at 25°C (literature))
Boiling Point 130°C to 135°C (1 mmHg)
Molecular Formula C15H27N3O
Refractive Index n20/D 1.516 (literature)
Linear Formula [(CH3)2 NCH2]3C6H2OH
MDL Number MFCD00008330
Quantity 50 mL
Synonym DMP-30
Molecular Weight (g/mol) 265.39
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.