Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine 95.0+%, TCI America™
SDP

Catalog No. T085801ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T085801ML 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T085801ML Supplier TCI America Supplier No. T08580.1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 858-46-8
Molecular Formula C9F15N3
Molecular Weight (g/mol) 435.10
MDL Number MFCD00042437
InChI Key MQBPAXAOMVELQG-UHFFFAOYSA-N
Synonym 2,4,6-Tris(perfluoroethyl)-1,3,5-triazine
PubChem CID 70072
IUPAC Name tris(1,1,2,2,2-pentafluoroethyl)-1,3,5-triazine
SMILES FC(F)(F)C(F)(F)C1=NC(=NC(=N1)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 121°C
Quantity 0.1 mL
Formula Weight 435.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.