Alfa Aesar™ 2,4,6-Tris(trifluoromethyl)-1,3,5-triazine, 98%

Manufacturer: Alfa Aesar™ B2200103

Quantity 1g

 View more versions of this product

Catalog No. AAB2200103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 368-66-1
    Molecular Formula C6F9N3
    Formula Weight 285.07
    MDL Number MFCD00042436
    Beilstein 302339
    Chemical Name or Material 2,4,6-Tris(trifluoromethyl)-1,3,5-triazine

    Specifications

    Formula Weight 285.07
    Molecular Formula C6F9N3
    MDL Number MFCD00042436
    CAS 368-66-1
    Beilstein 302339
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H332-H315-H319-H335
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 95° to 96°C
    Quantity 1g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number XZ2800000
    Chemical Name or Material 2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
    Assay 98%

    Description

    Applications
    2,4,6-Tris(trifluoromethyl)-1,3,5-triazine is used to prepare 1-phenyl-4,6-bis-trifluoromethyl-1H-pyrazolo[3,4-d]pyrimidine by reacting with sodium 5-amino-1-phenyl-1H-pyrazole-4-carboxylate.

    Notes
    Air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

    GHS H Statement
    H332-H315-H319-H335
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit