Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[[[4-(Acetylamino)phenyl]sulfonyl][(4-fluorophenyl)methyl]amino]-N-(9-ethyl-9H-carbazol-3-yl)acetamide | 785821-19-4 | 98.0% | 572.65 | 5 MG
SDP

Catalog No. 5000403817
Encompass_Preferred

This biological molecule is sold for research purposes only and not for patient use.

  • High purity of 98.0%
  • Molecular weight of 572.65
  • Chemical formula C31H29FN4O4S
  • Appears as a solid
  • Color is white to off-white
  • Ships at room temperature in continental US
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-03817 Supplier Medchemexpress LLC Supplier No. HY4022775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This biological molecule is sold for research purposes only and not for patient use.

  • High purity of 98.0%
  • Molecular weight of 572.65
  • Chemical formula C31H29FN4O4S
  • Appears as a solid
  • Color is white to off-white
  • Ships at room temperature in continental US
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.