Learn More
Medchemexpress LLC 2-((4-amino-1-(tert-butyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl)methyl)phenol | 833481-73-5 | MFCD28120704 | 99.7% | 297.35 g/mol | C16H19N5O | 1 ML
SDP
Catalog No. 5000248416
Encompass_Preferred
2OH-BNPP1 is a small-molecule inhibitor of BUB1 serine/threonine kinase used as a research reagent to study mitotic checkpoint signaling and cancer-related pathways. It is supplied in concentrated solution and solid formats for biochemical and cellular assays.
  • Inhibits BUB1 kinase activity for cell cycle and mitosis research.
  • Available as 10 mM solution in DMSO and as solid aliquots.
  • High purity suitable for biochemical and cellular experiments.
  • Provided with analytical and quality control data when available.
  • Suitable for inhibitor profiling, cell-based assays, and mechanistic studies.
Catalog No. 50-002-48416 Supplier Medchemexpress LLC Supplier No. HY10208110MM 1ML
2OH-BNPP1 is a small-molecule inhibitor of BUB1 serine/threonine kinase used as a research reagent to study mitotic checkpoint signaling and cancer-related pathways. It is supplied in concentrated solution and solid formats for biochemical and cellular assays.
  • Inhibits BUB1 kinase activity for cell cycle and mitosis research.
  • Available as 10 mM solution in DMSO and as solid aliquots.
  • High purity suitable for biochemical and cellular experiments.
  • Provided with analytical and quality control data when available.
  • Suitable for inhibitor profiling, cell-based assays, and mechanistic studies.