Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(4-amino-1-tert-butylpyrazolo[3,4-d]pyrimidin-3-yl)methyl]phenol | 833481-73-5 | MFCD28120704 | 99.7% | 297.35 | C16H19N5O | 25 MG
SDP

Catalog No. 5000248372
Encompass_Preferred

2OH-BNPP1 is a small-molecule inhibitor of the serine/threonine kinase BUB1, used as a research tool in oncology and cell-cycle studies. It is supplied as a white to off-white solid and as a DMSO solution, and is characterized by high reported purity and defined storage and solubility conditions.

  • Inhibitor of BUB1 kinase for research applications.
  • Reported purity 99.65% (analytical grade).
  • Molecular formula C16H19N5O; molecular weight 297.35 g/mol.
  • Physical form: white to off-white solid; also available as a DMSO solution.
  • Solubility: DMSO 100 mg/mL (336.30 mM); in vivo formulations and solubility notes provided.
  • Storage: powder stored at -20°C (long term) or 4°C; solutions recommended at -80°C for long-term storage.
  • Available in small research quantities, e.g., 25 mg, 50 mg, and 100 mg.

Catalog No. 50-002-48372 Supplier Medchemexpress LLC Supplier No. HY10208125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2OH-BNPP1 is a small-molecule inhibitor of the serine/threonine kinase BUB1, used as a research tool in oncology and cell-cycle studies. It is supplied as a white to off-white solid and as a DMSO solution, and is characterized by high reported purity and defined storage and solubility conditions.

  • Inhibitor of BUB1 kinase for research applications.
  • Reported purity 99.65% (analytical grade).
  • Molecular formula C16H19N5O; molecular weight 297.35 g/mol.
  • Physical form: white to off-white solid; also available as a DMSO solution.
  • Solubility: DMSO 100 mg/mL (336.30 mM); in vivo formulations and solubility notes provided.
  • Storage: powder stored at -20°C (long term) or 4°C; solutions recommended at -80°C for long-term storage.
  • Available in small research quantities, e.g., 25 mg, 50 mg, and 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.