Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(4-amino-1-tert-butylpyrazolo[3,4-d]pyrimidin-3-yl)methyl]phenol | 833481-73-5 | MFCD28120704 | 99.7% | 297.35 g/mol | C16H19N5O | 100 MG
SDP

Catalog No. 5000248369
Encompass_Preferred

2OH-BNPP1 is a small-molecule inhibitor of BUB1 kinase used for research into cell-cycle regulation and cancer-related kinase pathways. The compound is supplied as a white to off-white solid and is intended for in vitro biochemical and cellular assays.

  • Inhibits BUB1 serine/threonine kinase for studies of mitotic checkpoint signaling.
  • High purity (99.7%) appropriate for research applications.
  • Molecular formula C16H19N5O; molecular weight 297.35 g/mol.
  • Soluble in DMSO at ≥100 mg/mL; ultrasonic assistance recommended.
  • Supplied as a 100 mg solid and should be stored frozen for long-term stability.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-48369 Supplier Medchemexpress LLC Supplier No. HY102081100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2OH-BNPP1 is a small-molecule inhibitor of BUB1 kinase used for research into cell-cycle regulation and cancer-related kinase pathways. The compound is supplied as a white to off-white solid and is intended for in vitro biochemical and cellular assays.

  • Inhibits BUB1 serine/threonine kinase for studies of mitotic checkpoint signaling.
  • High purity (99.7%) appropriate for research applications.
  • Molecular formula C16H19N5O; molecular weight 297.35 g/mol.
  • Soluble in DMSO at ≥100 mg/mL; ultrasonic assistance recommended.
  • Supplied as a 100 mg solid and should be stored frozen for long-term stability.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.