Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(4-amino-1-tert-butylpyrazolo[3,4-d]pyrimidin-3-yl)methyl]phenol | 833481-73-5 | MFCD28120704 | 99.7% | 297.35 g/mol | C16H19N5O | 5 MG
SDP

Catalog No. 5000249062
Encompass_Preferred

2OH-BNPP1 is a research-grade small-molecule inhibitor of BUB1 (mitotic checkpoint serine/threonine kinase) intended for biochemical and cellular studies in cancer research. It is supplied as a white to off-white solid with high HPLC purity and is for research use only.

  • Inhibitor of BUB1 serine/threonine kinase.
  • Research use only; not for human or therapeutic use.
  • High purity: 99.7% HPLC.
  • Chemical formula C16H19N5O; molecular weight 297.35 g/mol.
  • Available in small milligram quantities for assay development and screening.
  • Recommended storage: powder at -20°C; store solutions at -80°C when applicable.

Catalog No. 50-002-49062 Supplier Medchemexpress LLC Supplier No. HY1020815MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2OH-BNPP1 is a research-grade small-molecule inhibitor of BUB1 (mitotic checkpoint serine/threonine kinase) intended for biochemical and cellular studies in cancer research. It is supplied as a white to off-white solid with high HPLC purity and is for research use only.

  • Inhibitor of BUB1 serine/threonine kinase.
  • Research use only; not for human or therapeutic use.
  • High purity: 99.7% HPLC.
  • Chemical formula C16H19N5O; molecular weight 297.35 g/mol.
  • Available in small milligram quantities for assay development and screening.
  • Recommended storage: powder at -20°C; store solutions at -80°C when applicable.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.