Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Aminophenyl)acetamidoxime, 97%, Thermo Scientific™

Catalog No. AAH66933MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66933MD 250 mg
AAH6693303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66933MD Supplier Thermo Scientific Chemicals Supplier No. H66933MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500587-42-8
Molecular Formula C8H11N3O
Molecular Weight (g/mol) 165.196
InChI Key BXIVTVQCXHGZMM-UHFFFAOYSA-N
Synonym 2-4-aminophenyl-n'-hydroxyethanimidamide
PubChem CID 9573319
IUPAC Name 2-(4-aminophenyl)-N'-hydroxyethanimidamide
SMILES C1=CC(=CC=C1CC(=NO)N)N

Specifications

Color Brown
Quantity 250 mg
Formula Weight 165.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(4-Aminophenyl)acetamidoxime
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.