Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Aminophenyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. A0394100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0394100G 100 g
A039425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0394100G Supplier TCI America Supplier No. A0394100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104-10-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007922
InChI Key QXHDYMUPPXAMPQ-UHFFFAOYSA-N
Synonym 2-4-aminophenyl ethanol, 4-aminophenethyl alcohol, p-aminophenethyl alcohol, p-aminophenylethanol, benzeneethanol, 4-amino, 2-p-aminophenyl ethanol, 2-4-aminophenyl ethan-1-ol, 4-2-hydroxyethyl aniline, benzeneethanol, ar-amino, p-2-hydroxyethyl aniline
PubChem CID 66904
IUPAC Name 2-(4-aminophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)N

Specifications

Melting Point 108°C
Boiling Point 255°C
Quantity 100 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Aminophenyl)ethanol
EINECSNumber (3)-1071
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.