Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-amylcinnamoyl)amino-4-chlorobenzoic acid | 99754-06-0 | 99.3% | 371.86 g/mol | C21H22ClNO3 | 5MG
SDP

Catalog No. 5000238107
Encompass_Preferred

ONO-RS-082 is a research-grade, cell-permeable small molecule inhibitor of Ca2+-independent phospholipase A2 (PLA2) used to study arachidonic acid metabolism and inflammatory signaling. It inhibits PLA2 with reported IC50 ≈ 1.0 μM, does not inhibit PLC at 100 μM, and has molecular weight 371.86 g/mol (C21H22ClNO3).

  • Reversible PLA2 inhibitor with IC50 ≈ 1.0 μM.
  • Does not inhibit PLC at concentrations up to 100 μM.
  • Molecular weight 371.86 g/mol.
  • Chemical formula C21H22ClNO3.
  • High purity (≈99.3%).
  • Supplied as solid and as 10 mM solution in DMSO.
  • Store powder at -20°C; store solutions at -80°C for long-term storage.

Catalog No. 50-002-38107 Supplier Medchemexpress LLC Supplier No. HY1230705MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ONO-RS-082 is a research-grade, cell-permeable small molecule inhibitor of Ca2+-independent phospholipase A2 (PLA2) used to study arachidonic acid metabolism and inflammatory signaling. It inhibits PLA2 with reported IC50 ≈ 1.0 μM, does not inhibit PLC at 100 μM, and has molecular weight 371.86 g/mol (C21H22ClNO3).

  • Reversible PLA2 inhibitor with IC50 ≈ 1.0 μM.
  • Does not inhibit PLC at concentrations up to 100 μM.
  • Molecular weight 371.86 g/mol.
  • Chemical formula C21H22ClNO3.
  • High purity (≈99.3%).
  • Supplied as solid and as 10 mM solution in DMSO.
  • Store powder at -20°C; store solutions at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.