Learn More
Medchemexpress LLC 2-((4-(Benzo[d]thiazol-5-ylamino)-6-(tert-butylsulfonyl)quinazolin-7-yl)oxy)ethyl Dihydrogen Phosphate | 99.5% | 612.63 | 50 MG
SDP
Catalog No. 5000439242
Encompass_Preferred
GSK2983559 is an orally active and potent receptor interacting protein 2 (RIP2) kinase inhibitor. It blocks many proinflammatory cytokine responses in vivo and in human inflammatory bowel disease explant samples.
  • Blocks MDP-induced IL-8 in THP-1 cells.
  • Inhibits MDP-induced IL-6 in mouse.
Catalog No. 50-004-39242 Supplier Medchemexpress LLC Supplier No. HY112038A50MG
GSK2983559 is an orally active and potent receptor interacting protein 2 (RIP2) kinase inhibitor. It blocks many proinflammatory cytokine responses in vivo and in human inflammatory bowel disease explant samples.
  • Blocks MDP-induced IL-8 in THP-1 cells.
  • Inhibits MDP-induced IL-6 in mouse.