Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole, 98+%, Thermo Scientific™

Catalog No. AAL0870103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0870103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0870103 Supplier Thermo Scientific Chemicals Supplier No. L0870103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21510-43-0
Molecular Formula C14H9BrN2O
Molecular Weight (g/mol) 301.14
MDL Number MFCD00050997
InChI Key CNLVYZSUMYQALH-UHFFFAOYSA-N
PubChem CID 88931
IUPAC Name 2-(4-bromophenyl)-5-phenyl-1,3,4-oxadiazole
SMILES BrC1=CC=C(C=C1)C1=NN=C(O1)C1=CC=CC=C1

Specifications

Melting Point 167°C to 172°C
Quantity 1 g
Beilstein 204891
Synonym 2-4-bromophenyl-5-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-bromophenyl-5-phenyl, 5-4-bromophenyl-2-phenyl-1,3,4-oxadiazole, acmc-1cq4f, cambridge id 5570574, 1,3,4-oxadiazole,2-4-bromophenyl-5-phenyl, 2-4-bromo-phenyl-5-phenyl-1,3,4 oxadiazo le
Formula Weight 301.14
Percent Purity ≥98%
Chemical Name or Material 2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole
EINECSNumber 244-413-6
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.