Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(4-bromophenyl)methylene]-N-(2,6-dimethylphenyl)-hydrazinecarboxamide | 415687-81-9 | 99.8% | 346.22 | C16H16BrN3O | 10MG
SDP

Catalog No. 5000237300
Encompass_Preferred

EGA is a small-molecule inhibitor that selectively disrupts trafficking from early to late endosomes, blocking the entry of acid-dependent bacterial toxins and viruses into mammalian cells. It is intended for research use to study endosomal trafficking, toxin and virus uptake, and receptor degradation pathways.

  • High purity (99.8%).
  • Molecular formula C16H16BrN3O.
  • Molecular weight 346.22 Da.
  • Soluble in DMSO.
  • Supplied in small research pack sizes (e.g., 10 MG).
  • Useful for cell biology studies of endosomal trafficking and toxin/virus entry.

Catalog No. 50-002-37300 Supplier Medchemexpress LLC Supplier No. HY12516810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGA is a small-molecule inhibitor that selectively disrupts trafficking from early to late endosomes, blocking the entry of acid-dependent bacterial toxins and viruses into mammalian cells. It is intended for research use to study endosomal trafficking, toxin and virus uptake, and receptor degradation pathways.

  • High purity (99.8%).
  • Molecular formula C16H16BrN3O.
  • Molecular weight 346.22 Da.
  • Soluble in DMSO.
  • Supplied in small research pack sizes (e.g., 10 MG).
  • Useful for cell biology studies of endosomal trafficking and toxin/virus entry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.