Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromophenyl)thiophene 97.0+%, TCI America™
SDP

Catalog No. B51065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51065G 5 g
B51061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51065G Supplier TCI America Supplier No. B51065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40133-22-0
Molecular Formula C10H7BrS
Molecular Weight (g/mol) 239.13
MDL Number MFCD05664340
InChI Key XKOJJKOSNQXQGP-UHFFFAOYSA-N
PubChem CID 7018298
IUPAC Name 2-(4-bromophenyl)thiophene
SMILES BrC1=CC=C(C=C1)C1=CC=CS1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 239.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.