Learn More
Medchemexpress LLC 2-(4-chloro-2-methylphenoxy)propanoic acid | 93-65-2 | MFCD00002648 | 99.8% | 214.65 g/mol | C10H11ClO3 | 250 MG
SDP
Catalog No. 5000228684
Encompass_Preferred
Mecoprop (2-(4-chloro-2-methylphenoxy)propanoic acid) is a phenoxy herbicide supplied as an analytical standard for research use. The product is provided as a solid 250 mg vial with high reported purity and supporting analytical documentation. It is intended for laboratory and analytical applications and is not for clinical or human use.
  • Analytical standard grade purity 99.8%.
  • Molecular weight 214.65 g/mol; formula C10H11ClO3.
  • Supplied as a 250 mg solid vial.
  • Includes datasheet, certificate of analysis, SDS, HNMR, and RP-HPLC documentation.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-28684 Supplier Medchemexpress LLC Supplier No. HY118126250MG
Mecoprop (2-(4-chloro-2-methylphenoxy)propanoic acid) is a phenoxy herbicide supplied as an analytical standard for research use. The product is provided as a solid 250 mg vial with high reported purity and supporting analytical documentation. It is intended for laboratory and analytical applications and is not for clinical or human use.
  • Analytical standard grade purity 99.8%.
  • Molecular weight 214.65 g/mol; formula C10H11ClO3.
  • Supplied as a 250 mg solid vial.
  • Includes datasheet, certificate of analysis, SDS, HNMR, and RP-HPLC documentation.
  • Intended for research use only; not for human or clinical use.