Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-Chloro-phenoxy)-phenylacetic acid | 25563-04-6 | 99.6% | 262.69 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY20152100MG

Encompass_Preferred

2-(4-Chloro-phenoxy)-phenylacetic acid is a drug intermediate used for the synthesis of various active compounds. This product is intended for research use only, and MedChemExpress does not sell to patients.

  • Suitable for high-speed applications
  • Room temperature shipping in continental US
  • Storage in powder form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-03940


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.