Learn More
Medchemexpress LLC 2-(4-Chloro-phenoxy)-phenylacetic acid | 25563-04-6 | 99.6% | 262.69 | 500 MG
Supplier: Medchemexpress LLC HY20152500MG
2-(4-Chloro-phenoxy)-phenylacetic acid is a chemical reagent used as a drug intermediate in the synthesis of various active compounds. It is intended for research use only and is not sold to patients. The product is shipped at room temperature within the continental US and should be stored as a powder at -20°C for up to 3 years or 4°C for up to 2 years. If in solvent, it should be stored at -80°C for 6 months or -20°C for 1 month.
- Drug intermediate for synthesis of various active compounds.
- For research use only.
- High purity (99.64%).
- Available in multiple pack sizes.