Learn More
Medchemexpress LLC 2-(4-chlorobenzyl)-3-hydroxy-7,8,9,10-tetrahydrobenzo[h]quinoline-4-carboxylic acid | 851546-61-7 | 99.8% | 367.83 g/mol | C21H18ClNO3 | 10 MG
SDP
Catalog No. 5000287485
Encompass_Preferred
PSI-697 is a small-molecule, orally active P-selectin inhibitor in the quinoline-carboxylic acid series that was investigated for antithrombotic and anti-inflammatory effects in preclinical models and early clinical studies.
  • Orally active small-molecule P-selectin inhibitor.
  • Characterized in vitro and in rodent models for antithrombotic activity.
  • Reported in vitro P-selectin inhibition in the 50-125 μM range.
  • High reported purity suitable for research use.
  • Available in multiple small pack sizes for laboratory studies.
Catalog No. 50-002-87485 Supplier Medchemexpress LLC Supplier No. HY1552610MG
PSI-697 is a small-molecule, orally active P-selectin inhibitor in the quinoline-carboxylic acid series that was investigated for antithrombotic and anti-inflammatory effects in preclinical models and early clinical studies.
  • Orally active small-molecule P-selectin inhibitor.
  • Characterized in vitro and in rodent models for antithrombotic activity.
  • Reported in vitro P-selectin inhibition in the 50-125 μM range.
  • High reported purity suitable for research use.
  • Available in multiple small pack sizes for laboratory studies.