Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(4-Chlorobutyl)-1,3-dioxolane

Catalog No. 240455MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-045-5MLF 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 24-045-5MLF Supplier Sigma Aldrich Supplier No. 240455MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(4-Chlorobutyl)-1,3-dioxolane

Specifications

CAS 118336-86-0
Density 1.109 g/mL (at 20°C (literature))
Boiling Point 56°C to 58°C (0.1 mmHg)
Molecular Formula C7H13ClO2
Refractive Index n20/D 1.457
Linear Formula C7H13ClO2
MDL Number MFCD00043136
Quantity 500 mg
Synonym 5-Chloro-n-valeraldehyde ethylene acetal
Molecular Weight (g/mol) 164.63
Percent Purity ≥97.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.