Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-chlorophenoxy)ethanol | 1892-43-9 | 97.3% | 172.61 g/mol | C8H9ClO2 | 1 G
SDP

Catalog No. 5000302020
Encompass_Preferred

2-(4-Chlorophenoxy)ethanol is a small-molecule intermediate used in organic synthesis, commonly in the preparation of herbicides and related compounds. Supplied as a solid with reported high purity, it is intended for research use and laboratory synthesis.

  • High purity (97.34%) suitable for research applications.
  • White to light yellow solid appearance.
  • Molecular weight 172.61 g/mol.
  • Chemical formula C8H9ClO2.
  • Soluble in DMSO (50 mg/mL; may require sonication).
  • Available in small research pack sizes; larger quantities available by quote.

Catalog No. 50-003-02020 Supplier Medchemexpress LLC Supplier No. HYW0168571G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(4-Chlorophenoxy)ethanol is a small-molecule intermediate used in organic synthesis, commonly in the preparation of herbicides and related compounds. Supplied as a solid with reported high purity, it is intended for research use and laboratory synthesis.

  • High purity (97.34%) suitable for research applications.
  • White to light yellow solid appearance.
  • Molecular weight 172.61 g/mol.
  • Chemical formula C8H9ClO2.
  • Soluble in DMSO (50 mg/mL; may require sonication).
  • Available in small research pack sizes; larger quantities available by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.