Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-chlorophenoxy)ethanol | 1892-43-9 | MFCD00014036 | 97.3% | 172.61 | C8H9ClO2 | 1 ML
SDP

Catalog No. 5000302019
Encompass_Preferred

2-(4-chlorophenoxy)ethanol is an aromatic ether alcohol used primarily as a chemical intermediate, for example in herbicide synthesis. It is supplied in solid form and as a ready-to-use 10 mM solution in DMSO for research applications.

  • Chemical formula C8H9ClO2 and molecular weight 172.61 g/mol.
  • Purity typically about 97.3% (GC) for supplied lots.
  • Appearance: white to light yellow solid.
  • Available as solid (1 g, 5 g) and 10 mM in DMSO (1 mL) solution.
  • Storage: powder at -20°C (up to 3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Used as an intermediate in agrochemical and related syntheses.

Catalog No. 50-003-02019 Supplier Medchemexpress LLC Supplier No. HYW01685710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(4-chlorophenoxy)ethanol is an aromatic ether alcohol used primarily as a chemical intermediate, for example in herbicide synthesis. It is supplied in solid form and as a ready-to-use 10 mM solution in DMSO for research applications.

  • Chemical formula C8H9ClO2 and molecular weight 172.61 g/mol.
  • Purity typically about 97.3% (GC) for supplied lots.
  • Appearance: white to light yellow solid.
  • Available as solid (1 g, 5 g) and 10 mM in DMSO (1 mL) solution.
  • Storage: powder at -20°C (up to 3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Used as an intermediate in agrochemical and related syntheses.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.