Learn More
Medchemexpress LLC 2-[(4-chlorophenyl)sulfanyl]-N-[4-(pyridin-2-yl)-1,3-thiazol-2-yl]acetamide | 895470-67-4 | 99.7% | 361.87 | C16H12ClN3OS2 | 25MG
SDP
Catalog No. 5000237110
Encompass_Preferred
CP-312 is a small-molecule inducer of heme oxygenase-1 (HO-1) intended for in vitro research applications. The compound is an off-white to pink solid with a reported purity of 99.69% and molecular formula C16H12ClN3OS2; it dissolves in DMSO at approximately 125 mg/mL (sonication recommended). Use appropriate laboratory safety and handling procedures.
Catalog No. 50-002-37110 Supplier Medchemexpress LLC Supplier No. HY12481525MG
CP-312 is a small-molecule inducer of heme oxygenase-1 (HO-1) intended for in vitro research applications. The compound is an off-white to pink solid with a reported purity of 99.69% and molecular formula C16H12ClN3OS2; it dissolves in DMSO at approximately 125 mg/mL (sonication recommended). Use appropriate laboratory safety and handling procedures.