Learn More
Medchemexpress LLC 2-(4-chlorophenyl)sulfanyl-N-4-(pyridin-2-yl)-1,3-thiazol-2-ylacetamide | 895470-67-4 | 99.7% | 361.87 g/mol | C16H12ClN3OS2 | 100MG
SDP
Catalog No. 5000239475
Encompass_Preferred
CP-312 is a small-molecule inducer of heme oxygenase-1 (HO-1) used in research to activate antioxidant response pathways and protect cells from oxidative stress. Supplied as a high-purity solid (CAS 895470-67-4), it is suitable for biochemical and cellular assays, including studies of HMOX1 induction and cytoprotection.
  • Induces heme oxygenase-1 and activates antioxidant response.
  • High purity suitable for cellular and biochemical assays.
  • Solid powder form with good solubility in DMSO.
  • Useful for cardiomyocyte protection and oxidative stress models.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-39475 Supplier Medchemexpress LLC Supplier No. HY124815100MG
CP-312 is a small-molecule inducer of heme oxygenase-1 (HO-1) used in research to activate antioxidant response pathways and protect cells from oxidative stress. Supplied as a high-purity solid (CAS 895470-67-4), it is suitable for biochemical and cellular assays, including studies of HMOX1 induction and cytoprotection.
  • Induces heme oxygenase-1 and activates antioxidant response.
  • High purity suitable for cellular and biochemical assays.
  • Solid powder form with good solubility in DMSO.
  • Useful for cardiomyocyte protection and oxidative stress models.
  • Available in multiple pack sizes for experimental flexibility.