Learn More
Medchemexpress LLC 2-(4-chlorophenyl)thiazol-4-yl acetic acid | 17969-20-9 | 99.5% | 253.70 | C11H8ClNO2S | 10 MG
SDP
Catalog No. 5000247882
Encompass_Preferred
Fenclozic acid is an orally active analgesic, antipyretic, and anti-inflammatory research compound commonly used in preclinical pharmacology and biochemical assays. It is supplied as a solid and as DMSO solutions for in vitro testing and analytical characterization.
  • High purity (LCMS 99.52%) for reliable experimental results.
  • Characterized by LCMS and supplied with a certificate of analysis.
  • Suitable for in vitro pharmacology and biochemical assays.
  • Available as milligram-scale quantities and as ready-to-use DMSO solutions.
  • Molecular formula C11H8ClNO2S and molecular weight 253.70 for reference.
Catalog No. 50-002-47882 Supplier Medchemexpress LLC Supplier No. HY10143210MG
Fenclozic acid is an orally active analgesic, antipyretic, and anti-inflammatory research compound commonly used in preclinical pharmacology and biochemical assays. It is supplied as a solid and as DMSO solutions for in vitro testing and analytical characterization.
  • High purity (LCMS 99.52%) for reliable experimental results.
  • Characterized by LCMS and supplied with a certificate of analysis.
  • Suitable for in vitro pharmacology and biochemical assays.
  • Available as milligram-scale quantities and as ready-to-use DMSO solutions.
  • Molecular formula C11H8ClNO2S and molecular weight 253.70 for reference.