Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane, AcroSeal™

Catalog No. AC428991000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428991000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428991000 Supplier Thermo Scientific Chemicals Supplier No. 428991000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 126519-89-9 | C11H17ClO5SSi | 324.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126519-89-9 , 75-09-2
Molecular Formula C11H17ClO5SSi
Molecular Weight (g/mol) 324.85
MDL Number MFCD00054774
InChI Key NYIDSUMRGUILGR-UHFFFAOYSA-N
Synonym 2-4-chlorosulfonylphenyl ethyltrimethoxysilane, 4-2-trimethoxysilylethyl benzenesulfonyl chloride, 4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride, benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl, 2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid, 4-2-trimethoxysilyl ethyl benzenesulfonyl chloride, acmc-20ete8, 4-chlorosulfonyl phenethyltrimethoxysilane
PubChem CID 2733807
IUPAC Name 4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
SMILES CO[Si](CCC1=CC=C(C=C1)S(Cl)(=O)=O)(OC)OC

Specifications

CAS Min % 45.0
CAS Max % 55.0
Color Yellow
Boiling Point >40.0°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Solubility Information Solubility in water: reacts
Formula Weight 324.85
Percent Purity 45 to 55% (solids)
Physical Form Solution
Chemical Name or Material 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.