Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[[4-(cyclopropylcarbonyl)-1-piperazinyl]methyl]-1,2-benzisothiazol-3(2H)-one | 877963-94-5 | MFCD07849042 | 98.0% | 317.41 g/mol | C16H19N3O2S | 100 MG
SDP

Catalog No. 5000215041
Encompass_Preferred

LOC14 is a small-molecule inhibitor of protein disulfide isomerase (PDI) used in biochemical and cellular research to probe PDI activity and related pathways. It is supplied as a white to off-white solid with available solubility and in vivo formulation guidance for experimental use.

  • Potent PDI inhibitor used to study disulfide isomerase activity.
  • High purity (98.0%).
  • White to off-white solid suitable for analytical and biological assays.
  • Molecular weight 317.41 g/mol; formula C16H19N3O2S.
  • Soluble in DMSO; in vivo formulations and solubility data available.
  • Store at 4°C and prepare solutions fresh due to instability in solution.

Catalog No. 50-002-15041 Supplier Medchemexpress LLC Supplier No. HY100432100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LOC14 is a small-molecule inhibitor of protein disulfide isomerase (PDI) used in biochemical and cellular research to probe PDI activity and related pathways. It is supplied as a white to off-white solid with available solubility and in vivo formulation guidance for experimental use.

  • Potent PDI inhibitor used to study disulfide isomerase activity.
  • High purity (98.0%).
  • White to off-white solid suitable for analytical and biological assays.
  • Molecular weight 317.41 g/mol; formula C16H19N3O2S.
  • Soluble in DMSO; in vivo formulations and solubility data available.
  • Store at 4°C and prepare solutions fresh due to instability in solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.