Learn More
Medchemexpress LLC 2,4-D Butyl ester | 94-80-4 | 99.9% | 1 ML
SDP
Catalog No. 5000408904
Encompass_Preferred
2,4-D Butyl ester is an organic phenoxy herbicide used to control woody broad-leaf weeds. It produces its herbicidal effect by mimicking natural plant growth hormones, causing plants to grow too rapidly to survive.
  • Organic phenoxy herbicide
  • Controls woody broad-leaf weeds
  • Mimics natural plant growth hormones
Catalog No. 50-004-08904 Supplier Medchemexpress LLC Supplier No. HYB086710MM/1ML
2,4-D Butyl ester is an organic phenoxy herbicide used to control woody broad-leaf weeds. It produces its herbicidal effect by mimicking natural plant growth hormones, causing plants to grow too rapidly to survive.
  • Organic phenoxy herbicide
  • Controls woody broad-leaf weeds
  • Mimics natural plant growth hormones