Learn More
Medchemexpress LLC 2,4-D isooctyl ester | 25168-26-7 | 97.2% | 10 MM 1 ML
SDP
Catalog No. 5000408862
Encompass_Preferred
2,4-D isooctyl ester is a selective herbicide of the chlorophenoxy compound family, known for its higher boiling point, low volatility, and minimal drift. It is effective in managing broadleaf weeds across various environments, including crops, lawns, and forests.
  • Selective herbicide
  • Chlorophenoxy compound
  • Higher boiling point
  • Low volatility
  • Minimal drift
  • Utilized to manage broadleaf weeds
  • Suitable for use in crops, lawns, and forests
Catalog No. 50-004-08862 Supplier Medchemexpress LLC Supplier No. HYB086810MM/1ML
2,4-D isooctyl ester is a selective herbicide of the chlorophenoxy compound family, known for its higher boiling point, low volatility, and minimal drift. It is effective in managing broadleaf weeds across various environments, including crops, lawns, and forests.
  • Selective herbicide
  • Chlorophenoxy compound
  • Higher boiling point
  • Low volatility
  • Minimal drift
  • Utilized to manage broadleaf weeds
  • Suitable for use in crops, lawns, and forests