Learn More
Medchemexpress LLC 2,4-D methyl ester | 1928-38-7 | MFCD00018356 | >98.0% | 235.06 g/mol | C9H8Cl2O3 | 5G
SDP
Catalog No. 5000238954
Encompass_Preferred
Methyl 2,4-dichlorophenoxyacetate (2,4-D methyl ester, CAS 1928-38-7) is the methyl ester derivative of the herbicide 2,4-dichlorophenoxyacetic acid supplied as a research-grade analytical chemical. It is a solid organic compound with formula C9H8Cl2O3 and molecular weight 235.06 g/mol, specified at ≥98.0% purity and offered in small laboratory pack sizes. Follow product label for storage and handling recommendations.
  • High purity ≥98.0% suitable for analytical applications.
  • Available in small pack sizes for lab-scale use.
  • Solid form simplifies handling and storage.
  • Well-defined chemical identity (C9H8Cl2O3; MW 235.06 g/mol).
  • Recommended solvent storage conditions for extended stability.
Catalog No. 50-002-38954 Supplier Medchemexpress LLC Supplier No. HY1245475G
Methyl 2,4-dichlorophenoxyacetate (2,4-D methyl ester, CAS 1928-38-7) is the methyl ester derivative of the herbicide 2,4-dichlorophenoxyacetic acid supplied as a research-grade analytical chemical. It is a solid organic compound with formula C9H8Cl2O3 and molecular weight 235.06 g/mol, specified at ≥98.0% purity and offered in small laboratory pack sizes. Follow product label for storage and handling recommendations.
  • High purity ≥98.0% suitable for analytical applications.
  • Available in small pack sizes for lab-scale use.
  • Solid form simplifies handling and storage.
  • Well-defined chemical identity (C9H8Cl2O3; MW 235.06 g/mol).
  • Recommended solvent storage conditions for extended stability.