Learn More
Medchemexpress LLC 2,4-D sodium (Standard) | 2702-72-9 | 97% | 243.02 | 100 MG
SDP
Catalog No. 5000387857
Encompass_Preferred
2,4-D sodium (Standard) is an analytical standard intended for research and analytical applications. It functions as a selective herbicide, controlling broad-leaved weeds. The compound induces apoptosis and inhibits DNA and protein synthesis, thereby preventing normal plant growth and development. It serves as a reference standard for assay.
  • Intended for research and analytical applications.
  • Selective herbicide for broad-leaved weeds.
  • Induces apoptosis.
  • Inhibits DNA and protein synthesis.
  • Prevents normal plant growth and development.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-003-87857 Supplier Medchemexpress LLC Supplier No. HY18572AR100MG
2,4-D sodium (Standard) is an analytical standard intended for research and analytical applications. It functions as a selective herbicide, controlling broad-leaved weeds. The compound induces apoptosis and inhibits DNA and protein synthesis, thereby preventing normal plant growth and development. It serves as a reference standard for assay.
  • Intended for research and analytical applications.
  • Selective herbicide for broad-leaved weeds.
  • Induces apoptosis.
  • Inhibits DNA and protein synthesis.
  • Prevents normal plant growth and development.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.