Learn More
Crescent Chemical Co Inc 2,4 -DDE SOLN 1ML
SDP
Catalog No. 5070749
Encompass_Preferred
2,4'-DDE Single Component Calibration Standard in Methanol CAS 3424-82-6 Size - 1mL Catalog Number - 7347M
Catalog No. 50-707-49 Supplier Crescent Chemical Co Inc Supplier No. CT7347M
2,4'-DDE Single Component Calibration Standard in Methanol CAS 3424-82-6 Size - 1mL Catalog Number - 7347M