Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Decadien-1-ol 95.0+%, TCI America™
SDP

Catalog No. D19355ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19355ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D19355ML Supplier TCI America Supplier No. D19355ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14507-02-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00014052
InChI Key NUBWFSDCZULDCI-BLHCBFLLSA-N
PubChem CID 5362696
IUPAC Name (2E,4E)-deca-2,4-dien-1-ol
SMILES CCCCCC=CC=CCO

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Decadien-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.