Learn More
Apexbio Technology LLC 2,4-Di-tert-butylphenol 96-76-4 1g
SDP
Catalog No. 5000089681
Encompass_Preferred
2 4-Di-tert-butylphenol (CAS 96-76-4) is an endogenous metabolic compound recognized for its antioxidant and anti-inflammatory properties Mechanistic studies have demonstrated that 2 4-Di-tert-butylphenol modulates the expression of genes involved in apoptotic pathways leading to the induction of apoptosis in cancer cell models Additionally experimental evidence indicates antiviral and antifungal activities supporting its use in investigations of infection mechanisms This compound is therefore relevant for research in oncology inflammation and the study of pathogen-host interactions
Catalog No. 50-000-89681 Supplier Apexbio Technology LLC Supplier No. C68681000
2 4-Di-tert-butylphenol (CAS 96-76-4) is an endogenous metabolic compound recognized for its antioxidant and anti-inflammatory properties Mechanistic studies have demonstrated that 2 4-Di-tert-butylphenol modulates the expression of genes involved in apoptotic pathways leading to the induction of apoptosis in cancer cell models Additionally experimental evidence indicates antiviral and antifungal activities supporting its use in investigations of infection mechanisms This compound is therefore relevant for research in oncology inflammation and the study of pathogen-host interactions