Learn More
Medchemexpress LLC 2,4-di-tert-butylphenol | 96-76-4 | MFCD00008828 | 100.0% | 206.33 g/mol | C14H22O | 1 ML
SDP
Catalog No. 5000302914
Encompass_Preferred
2,4-di-tert-butylphenol is a substituted phenol supplied as a 10 mM solution in DMSO for research and analytical applications. The compound has a molecular weight of 206.33 g/mol, a molecular formula of C14H22O, and high reported purity, with supporting COA and SDS documents available from the manufacturer.
  • High purity suitable for analytical and research use.
  • Provided as a ready-to-use 10 mM solution in DMSO for convenience.
  • Small 1 mL volume ideal for screening and small-scale experiments.
  • Accompanied by certificate of analysis and safety data sheet for traceability.
  • Stable storage and handling information available in SDS.
Catalog No. 50-003-02914 Supplier Medchemexpress LLC Supplier No. HYW01458910MM/1ML
2,4-di-tert-butylphenol is a substituted phenol supplied as a 10 mM solution in DMSO for research and analytical applications. The compound has a molecular weight of 206.33 g/mol, a molecular formula of C14H22O, and high reported purity, with supporting COA and SDS documents available from the manufacturer.
  • High purity suitable for analytical and research use.
  • Provided as a ready-to-use 10 mM solution in DMSO for convenience.
  • Small 1 mL volume ideal for screening and small-scale experiments.
  • Accompanied by certificate of analysis and safety data sheet for traceability.
  • Stable storage and handling information available in SDS.