Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-di-tert-butylphenol | 96-76-4 | MFCD00008828 | 99.7% | 206.32 g/mol | C14H22O | 250 MG
SDP

Catalog No. 5000303621
Encompass_Preferred

2,4-Di-tert-butylphenol is a high-purity phenolic standard used as a reference compound in analytical and biochemical research. It has reported RXRα activity and antioxidant and anti-inflammatory properties, and is supplied as a characterized material for assay development and method validation.

  • Purity: 99.7%.
  • Pack size: 250 mg.
  • Molecular formula: C14H22O.
  • Molecular weight: 206.32 g/mol.
  • CAS number: 96-76-4.
  • Applications: reference standard for analytical assays and biochemical research.

Catalog No. 50-003-03621 Supplier Medchemexpress LLC Supplier No. 250MGHYW014589R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4-Di-tert-butylphenol is a high-purity phenolic standard used as a reference compound in analytical and biochemical research. It has reported RXRα activity and antioxidant and anti-inflammatory properties, and is supplied as a characterized material for assay development and method validation.

  • Purity: 99.7%.
  • Pack size: 250 mg.
  • Molecular formula: C14H22O.
  • Molecular weight: 206.32 g/mol.
  • CAS number: 96-76-4.
  • Applications: reference standard for analytical assays and biochemical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.