Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-diethylamino-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D051225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D051225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D051225G Supplier TCI America Supplier No. D051225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2073-31-6
Molecular Formula C7H14N6
Molecular Weight (g/mol) 182.23
MDL Number MFCD00059765
InChI Key LLOVZIWCKUWRTA-UHFFFAOYSA-N
Synonym N2,N2-Diethylmelamine
PubChem CID 326489
IUPAC Name N2,N2-diethyl-1,3,5-triazine-2,4,6-triamine
SMILES CCN(CC)C1=NC(N)=NC(N)=N1

Specifications

Melting Point 169°C
Color White
Quantity 25 g
Formula Weight 182.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-diethylamino-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.