Learn More
Medchemexpress LLC HY-100954 100mg Medchemexpress, 2,4-Diamino-6-hydroxypyrimidine CAS:56-06-4 Purity:>98%
SDP
Catalog No. 501967792
Encompass_Preferred
Medchemexpress, HY-100954 100mg 2,4-Diamino-6-hydroxypyrimidine CAS:56-06-4 2,4-Diamino-6-hydroxypyrimidine is a specific GTP cyclohydrolase I inhibitor (the rate-limiting enzyme in de novo pterin synthesis), blocks BH4 synthesis and suppresses NO production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7792 Supplier Medchemexpress LLC Supplier No. HY100954100MG
Medchemexpress, HY-100954 100mg 2,4-Diamino-6-hydroxypyrimidine CAS:56-06-4 2,4-Diamino-6-hydroxypyrimidine is a specific GTP cyclohydrolase I inhibitor (the rate-limiting enzyme in de novo pterin synthesis), blocks BH4 synthesis and suppresses NO production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.