Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-diamino-6-hydroxypyrimidine | 56-06-4 | MFCD00006098 | >98.0% | 126.12 g/mol | C4H6N4O | 1 G
SDP

Supplier:  Medchemexpress LLC HY1009541G

Encompass_Preferred

2,4-diamino-6-hydroxypyrimidine is a small-molecule research reagent used primarily as an inhibitor of GTP cyclohydrolase I (GCH1). It is supplied as a solid and is used in biochemical and enzymatic studies to probe folate and pteridine biosynthesis pathways.

  • acts as a GTP cyclohydrolase I (GCH1) inhibitor
  • molecular weight 126.12 g/mol
  • CAS number 56-06-4
  • purity greater than 98.0% (HPLC)
  • appearance off-white to light yellow solid
  • suitable for enzymatic assays and biochemical research

Catalog No. 50-002-12246


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.