Learn More
Medchemexpress LLC 2,4-diamino-6-hydroxypyrimidine | 56-06-4 | MFCD00006098 | 99.9% | 126.12 g/mol | C4H6N4O | 1 ML
SDP
Catalog No. 5000212270
Encompass_Preferred
2,4-diamino-6-hydroxypyrimidine is a small-molecule research reagent that acts as a specific inhibitor of GTP cyclohydrolase I, used to modulate tetrahydrobiopterin (BH4) synthesis and nitric oxide production in biochemical and cellular assays. The compound is supplied as a solid and as a pre-made 10 mM solution in DMSO, and batch documentation is available.
Catalog No. 50-002-12270 Supplier Medchemexpress LLC Supplier No. HY10095410MM 1ML
2,4-diamino-6-hydroxypyrimidine is a small-molecule research reagent that acts as a specific inhibitor of GTP cyclohydrolase I, used to modulate tetrahydrobiopterin (BH4) synthesis and nitric oxide production in biochemical and cellular assays. The compound is supplied as a solid and as a pre-made 10 mM solution in DMSO, and batch documentation is available.