Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diaminoanisole, MP Biomedicals

Catalog No. ICN212669 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212669 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212669 Supplier MP Biomedicals, Inc Supplier No. 212669

Chemical Identifiers

CAS 615-05-4
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
InChI Key BAHPQISAXRFLCL-UHFFFAOYSA-N
Synonym 2,4-diaminoanisole, 2,4-diaminoanisol, 1,3-benzenediamine, 4-methoxy, m-diaminoanisole, pelagol l, pelagol da, pelagol grey l, 4-methoxy-m-phenylenediamine, furro l, 2,4-diamineanisole
PubChem CID 11976
ChEBI CHEBI:82309
IUPAC Name 4-methoxybenzene-1,3-diamine
SMILES COC1=C(C=C(C=C1)N)N

Specifications

Quantity 100 g
Formula Weight 138.17
Chemical Name or Material 2, 4-Diaminoanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.