Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diaminoanisole Sulfate Hydrate 98.0+%, TCI America™
SDP

Catalog No. D130925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D130925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D130925G Supplier TCI America Supplier No. D130925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6219-67-6
Molecular Formula C7H12N2O5S
Molecular Weight (g/mol) 236.24
MDL Number MFCD00012978
InChI Key FDJQHYWUHGBBTO-UHFFFAOYSA-N
Synonym 4-Methoxy-1,3-phenylenediamine Sulfate
IUPAC Name 4-methoxybenzene-1,3-diamine; sulfuric acid
SMILES OS(O)(=O)=O.COC1=CC=C(N)C=C1N

Specifications

Quantity 25 g
Formula Weight 236.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diaminoanisole Sulfate Hydrate
EINECSNumber (5)-3031
RTECSNumber ST2705000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.